Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029983.2 | NP_001025154.1 | 2 | 264 | ||
GenBank mRNA | AK043541.1 | - | 2 | 245 | ||
BC138550.1 | - | 2 | 220 | 8861570 | ||
BC138551.1 | - | 2 | 220 | 8861571 | ||
BY732333.1 | - | 2 | 260 |