Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029983.2 | NP_001025154.1 | 5 | 652 | ||
GenBank mRNA | AK043541.1 | - | 5 | 633 | ||
AK178205.1 | - | 1 | 35 | |||
BC006645.1 | - | 3 | 268 | |||
BC138550.1 | - | 5 | 608 | 8861570 | ||
BC138551.1 | - | 5 | 608 | 8861571 | ||
BY732333.1 | - | 5 | 648 |