Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qsox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042036.1 | - | 3 | 444 | ||
AK140324.1 | - | 6 | 822 | |||
AK157992.1 | - | 6 | 815 | |||
AK165642.1 | - | 6 | 824 | |||
BC030934.1 | - | 6 | 856 | 4920915 |