Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011767.2 | NP_035897.2 | 20 | 3162 | ||
GenBank mRNA | AF071059.1 | - | 20 | 3160 | ||
AK011329.1 | - | 5 | 482 | |||
AK034963.1 | - | 13 | 1731 | |||
AK041429.1 | - | 4 | 415 | |||
AK158938.1 | - | 12 | 1470 | |||
BC006962.1 | - | 10 | 1209 | |||
BC094220.1 | - | 19 | 3049 | |||
BC141139.1 | - | 20 | 3146 | 9055751 |