Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011767.2 | NP_035897.2 | 19 | 3080 | ||
GenBank mRNA | AF071059.1 | - | 19 | 3078 | ||
AK011329.1 | - | 4 | 400 | |||
AK034963.1 | - | 12 | 1649 | |||
AK041429.1 | - | 3 | 333 | |||
AK158938.1 | - | 11 | 1388 | |||
BC006962.1 | - | 9 | 1127 | |||
BC094220.1 | - | 18 | 2967 | |||
BC141139.1 | - | 19 | 3064 | 9055751 |