Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290503.1 | NP_001277432.1 | 7 | 1552 | ||
NM_178694.4 | NP_848809.2 | 7 | 1591 | |||
GenBank mRNA | AK029290.1 | - | 7 | 1526 | ||
AK032153.1 | - | 6 | 1620 | |||
AK035712.1 | - | 3 | 2373 | |||
AK048853.1 | - | 7 | 1447 | |||
AK081355.1 | - | 7 | 1429 | |||
AK087046.1 | - | 7 | 1425 | |||
AK155401.1 | - | 7 | 1499 | |||
BC048924.1 | - | 7 | 1434 | 6312691 | ||
BC057021.1 | - | 7 | 1496 | 6825937 |