Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290503.1 | NP_001277432.1 | 14 | 2501 | ||
NM_178694.4 | NP_848809.2 | 14 | 2540 | |||
GenBank mRNA | AK155401.1 | - | 14 | 2448 | ||
BC048924.1 | - | 14 | 2383 | 6312691 | ||
BC057021.1 | - | 14 | 2445 | 6825937 |