Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146119.2 | NP_666231.1 | 3 | 504 | ||
GenBank mRNA | AK033735.1 | - | 3 | 472 | ||
AK046583.1 | - | 3 | 401 | |||
AK051396.1 | - | 3 | 443 | |||
AK146953.1 | - | 3 | 489 | |||
AK149818.1 | - | 3 | 415 | |||
AK149888.1 | - | 3 | 414 | |||
AK150642.1 | - | 3 | 442 | |||
AK152207.1 | - | 3 | 463 | |||
AK153489.1 | - | 3 | 448 | |||
AK170512.1 | - | 3 | 444 | |||
BC027843.1 | - | 3 | 424 | 5343013 |