Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146119.2 | NP_666231.1 | 9 | 1332 | ||
GenBank mRNA | AK033735.1 | - | 9 | 1300 | ||
AK046583.1 | - | 9 | 1229 | |||
AK051396.1 | - | 9 | 1271 | |||
AK146953.1 | - | 9 | 1317 | |||
AK149818.1 | - | 9 | 1243 | |||
AK149888.1 | - | 9 | 1242 | |||
AK150642.1 | - | 9 | 1270 | |||
AK152207.1 | - | 9 | 1291 | |||
AK153489.1 | - | 9 | 1276 | |||
AK170512.1 | - | 9 | 1272 | |||
BC024639.1 | - | 1 | 65 | |||
BC027843.1 | - | 9 | 1252 | 5343013 |