Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145523.3 | NP_663498.1 | 2 | 273 | ||
GenBank mRNA | AB088388.1 | - | 2 | 167 | ||
AF518325.1 | - | 2 | 257 | |||
AK033740.1 | - | 2 | 306 | |||
AK046347.1 | - | 2 | 315 | |||
AK049317.1 | - | 2 | 274 | |||
AK050540.1 | - | 2 | 315 | |||
AK078915.1 | - | 2 | 274 | |||
BC021450.1 | - | 2 | 267 | 5044040 |