Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172674.2 | NP_766262.2 | 14 | 2267 | ||
GenBank mRNA | AK033017.1 | - | 13 | 2066 | ||
AK190572.1 | - | 1 | 15 | |||
BC011337.1 | - | 2 | 226 | |||
BC060121.1 | - | 13 | 2186 | 6413021 |