Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144500.3 | NP_653083.2 | 10 | 1133 | ||
GenBank mRNA | AK048537.1 | - | 10 | 1053 | ||
AK145292.1 | - | 10 | 1165 | |||
AK161695.1 | - | 10 | 1072 | |||
AK166059.1 | - | 10 | 1112 | |||
AK170306.1 | - | 10 | 1117 | |||
AK170669.1 | - | 10 | 1134 | |||
AK178638.1 | - | 4 | 312 | |||
AK178810.1 | - | 4 | 363 | |||
AK182769.1 | - | 4 | 321 | |||
AK220257.1 | - | 4 | 452 | |||
BC026804.1 | - | 10 | 1140 | 4159217 |