Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zgpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048148.1 | NP_001041613.1 | 1 | 684 | ||
NM_144894.3 | NP_659143.1 | 2 | 908 | |||
GenBank mRNA | AK041491.1 | - | 2 | 912 | ||
AK049764.1 | - | 1 | 683 | |||
AK052236.1 | - | 1 | 582 | |||
AK159801.1 | - | 1 | 703 | |||
BC021513.1 | - | 1 | 682 | 5351756 | ||
BC027218.1 | - | 1 | 591 | 3158198 |