Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144987.1 | NP_001138459.1 | 4 | 1490 | ||
NM_144895.2 | NP_659144.1 | 4 | 1274 | |||
GenBank mRNA | AK031545.1 | - | 3 | 829 | ||
AK087388.1 | - | 4 | 1243 | |||
AK089946.1 | - | 4 | 1275 | |||
AK147760.1 | - | 4 | 1492 | |||
AK159951.1 | - | 4 | 1275 | |||
AK192376.1 | - | 2 | 176 | |||
BC022921.1 | - | 4 | 1242 | 4220506 |