Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177346.4 | NP_796320.2 | 4 | 2399 | ||
GenBank mRNA | AK035931.1 | - | 4 | 962 | ||
AK048373.1 | - | 1122 | ||||
AK136826.1 | - | 4 | 2400 | |||
AK166029.1 | - | 1227 | ||||
BC119599.2 | - | 4 | 1063 |
Set Membership: |
G-protein coupled receptor |