Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144896.4 | NP_659145.1 | 3 | 392 | ||
GenBank mRNA | AK028193.1 | - | 3 | 391 | ||
AK034702.1 | - | 3 | 393 | |||
AK053978.1 | - | 3 | 395 | |||
AK081800.1 | - | 3 | 396 | |||
AK134831.1 | - | 3 | 327 | |||
AK166231.1 | - | 3 | 403 | |||
BC005709.1 | - | 3 | 394 | 3157973 |