Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201407.4 | NP_958809.3 | 20 | 3452 | ||
GenBank mRNA | AK122294.1 | - | 7 | 2299 | ||
AK155638.1 | - | 20 | 3434 | |||
AY512564.1 | - | 19 | 3024 | |||
BC026996.1 | - | 1 | 302 | |||
BC040460.1 | - | 9 | 1499 | |||
BC049972.1 | - | 10 | 1738 | |||
BC066102.1 | - | 18 | 2886 | 30531856 | ||
BC151194.1 | - | 19 | 3098 | 9088082 |