Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177545.5 | NP_808213.2 | 5 | 1230 | ||
GenBank mRNA | AF481860.1 | - | 5 | 1146 | ||
AK135230.1 | - | 5 | 1191 | |||
AK167743.1 | - | 6 | 1044 | |||
BC138519.1 | - | 4 | 972 | 8861539 | ||
BC138520.1 | - | 4 | 972 | 8861540 | ||
BU610139.1 | - | 1 | 76 |