Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293689.1 | NP_001280618.1 | 4 | 989 | ||
NM_172271.2 | NP_758475.1 | 4 | 989 | |||
GenBank mRNA | AK032262.1 | - | 3 | 366 | ||
AK083807.1 | - | 4 | 890 | |||
AK085620.1 | - | 4 | 723 | |||
AK163113.1 | - | 4 | 993 | |||
AY155578.1 | - | 4 | 990 | |||
BC033610.1 | - | 4 | 614 | |||
BC141148.1 | - | 3 | 560 | 9055760 | ||
BC171962.1 | - | 3 | 560 | 9054248 |