Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286750.1 | NP_001273679.1 | 6 | 975 | ||
NM_153157.3 | NP_694797.1 | 6 | 1126 | |||
NM_153458.3 | NP_703188.1 | 6 | 1379 | |||
GenBank mRNA | AF442824.1 | - | 6 | 1126 | ||
AF442825.1 | - | 6 | 1094 | |||
AK038569.1 | - | 6 | 1326 | |||
AK044325.1 | - | 6 | 1181 | |||
AK044605.1 | - | 6 | 1329 | |||
AK045482.1 | - | 6 | 1328 | |||
AK049612.1 | - | 6 | 1380 | |||
AK079315.1 | - | 6 | 1189 | |||
AK137414.1 | - | 6 | 1181 | |||
BC112422.1 | - | 6 | 1035 | 40061966 | ||
BC113129.1 | - | 6 | 1035 | 40061967 |