Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144902.3 | NP_659151.1 | 6 | 943 | ||
GenBank mRNA | AK029398.1 | - | 6 | 1315 | ||
AK041195.1 | - | 7 | 1063 | |||
AK049484.1 | - | 6 | 943 | |||
AK161317.1 | - | 6 | 901 | |||
BC024110.1 | - | 6 | 939 | 5150304 |