Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293560.1 | NP_001280489.1 | 11 | 1191 | ||
NM_173763.4 | NP_776124.1 | 11 | 1195 | |||
GenBank mRNA | AF363737.1 | - | 11 | 1165 | ||
AK049569.1 | - | 11 | 1164 | |||
AK145623.1 | - | 11 | 1189 | |||
BC037135.1 | - | 7 | 772 | |||
BC085287.1 | - | 12 | 1288 | |||
BC131942.1 | - | 10 | 980 | 40130219 | ||
BC132615.1 | - | 10 | 980 | 40130892 |