Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139305.2 | NP_647466.2 | 8, 9 | 1166 | ||
GenBank mRNA | AJ245857.1 | - | 8, 9 | 1168 | ||
AK136579.1 | - | 8, 9 | 1167 | |||
AK137719.1 | - | 8, 9 | 1168 | |||
AK166403.1 | - | 5, 6 | 595 | |||
BC120544.1 | - | 8, 9 | 1159 | 40129467 | ||
BC120546.1 | - | 8, 9 | 1159 | 40129469 |