Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033306.1 | NP_001028478.1 | 4, 5 | 1769 | ||
GenBank mRNA | AK145414.1 | - | 4, 5 | 628 | ||
BC028832.1 | - | 3, 4 | 631 | |||
BC048850.1 | - | 3, 4 | 636 | |||
BC058986.1 | - | 4, 5 | 1271 |