Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172693.3 | NP_766281.1 | 2 | 461 | ||
GenBank mRNA | AB175680.1 | - | 2 | 441 | ||
AK033638.1 | - | 2 | 462 | |||
AK042133.1 | - | 2 | 494 | |||
AK082014.1 | - | 2 | 507 | |||
AK156751.1 | - | 2 | 491 | |||
BC025639.1 | - | 1 | 183 | |||
BC056425.1 | - | 2 | 472 | 5716909 |