Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173400.2 | NP_775576.2 | 8 | 1017 | ||
GenBank mRNA | AK051016.1 | - | 8 | 1038 | ||
AK077681.1 | - | 8 | 1039 | |||
AK169222.1 | - | 8 | 1019 | |||
AK173206.1 | - | 8 | 989 | |||
BC064725.1 | - | 3 | 214 | |||
BC068237.1 | - | 8 | 1017 | 30550558 |