Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146150.2 | NP_666262.2 | 30 | 3290 | ||
GenBank mRNA | AK031548.1 | - | 30 | 3290 | ||
AK050944.1 | - | 15 | 1389 | |||
AK166470.1 | - | 28 | 3289 | |||
BC023786.1 | - | 29 | 2668 | |||
BC024544.1 | - | 14 | 1275 | |||
BC026832.1 | - | 30 | 3340 | 4486176 | ||
BC029977.1 | - | 21 | 1926 | |||
BC036128.1 | - | 30 | 3408 | 4971791 |