Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146150.2 | NP_666262.2 | 27 | 2971 | ||
GenBank mRNA | AK031548.1 | - | 27 | 2971 | ||
AK166470.1 | - | 25 | 2970 | |||
BC023786.1 | - | 26 | 2349 | |||
BC024544.1 | - | 11 | 956 | |||
BC026832.1 | - | 27 | 3021 | 4486176 | ||
BC029977.1 | - | 18 | 1607 | |||
BC036128.1 | - | 27 | 3089 | 4971791 |