Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177669.4 | NP_808337.2 | 2, 3 | 232 | ||
GenBank mRNA | AK042513.1 | - | 2, 3 | 233 |