Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172698.2 | NP_766286.2 | 12 | 2064 | ||
GenBank mRNA | AK085129.1 | - | 1 | 640 | ||
AK132525.1 | - | 12 | 2365 | |||
AK163820.1 | - | 1 | 932 | |||
BC059213.1 | - | 12 | 2064 | 6408754 |