Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290696.1 | NP_001277625.1 | 6 | 836 | ||
NM_172699.3 | NP_766287.1 | 7 | 938 | |||
GenBank mRNA | AK082840.1 | - | 7 | 939 | ||
AK129271.1 | - | 6 | 809 | |||
AK139508.1 | - | 7 | 872 | |||
BC058231.1 | - | 7 | 911 | 6314981 | ||
CB245622.1 | - | 4 | 517 |