Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145552.2 | NP_663527.2 | 3 | 340 | ||
GenBank mRNA | AK037796.1 | - | 3 | 276 | ||
AK077532.1 | - | 3 | 310 | |||
AK146404.1 | - | 3 | 337 | |||
BC003262.1 | - | 3 | 316 | 3501393 | ||
BY140435.1 | - | 3 | 341 | |||
U69600.1 | - | 3 | 235 |