Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153159.2 | NP_694799.1 | 5 | 1020 | ||
GenBank mRNA | AK142501.1 | - | 5 | 1012 | ||
AK149698.1 | - | 5 | 1019 | |||
AK152196.1 | - | 5 | 977 | |||
AK152518.1 | - | 3 | 556 | |||
AK154363.1 | - | 5 | 993 | |||
AK161150.1 | - | 5 | 976 | |||
AK161733.1 | - | 5 | 975 | |||
AK172357.1 | - | 5 | 977 | |||
AY920404.1 | - | 4 | 871 | |||
BC006817.1 | - | 2 | 175 | |||
BC036563.1 | - | 5 | 979 | 5254681 |