Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thrap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146153.3 | NP_666265.3 | 4 | 1476 | ||
GenBank mRNA | AK034044.1 | - | 4 | 1356 | ||
AK079601.1 | - | 4 | 1356 | |||
BC012655.1 | - | 1 | 162 | |||
BC040346.1 | - | 4 | 1407 | |||
BC092239.1 | - | 4 | 1392 | 30659321 |