Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146155.3 | NP_666267.3 | 6 | 2879 | ||
GenBank mRNA | AK083440.1 | - | 1 | 129 | ||
AK134372.1 | - | 1 | 432 | |||
BC042621.1 | - | 6 | 2868 | 4911638 | ||
BC060231.1 | - | 5 | 2847 | 6852595 |