Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207237.3 | NP_997120.1 | 10 | 2348 | ||
GenBank mRNA | AK048880.1 | - | 5 | 590 | ||
AK144486.1 | - | 9 | 2122 | |||
BC030443.1 | - | 4 | 511 | |||
BC067023.1 | - | 11 | 1918 | 6848230 |