Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207237.3 | NP_997120.1 | 12 | 2667 | ||
GenBank mRNA | AK048880.1 | - | 7 | 909 | ||
AK144486.1 | - | 11 | 2441 | |||
AK191675.1 | - | 1 | 65 | |||
AK197093.1 | - | 1 | 65 | |||
BC030443.1 | - | 6 | 830 | |||
BC067023.1 | - | 13 | 2237 | 6848230 |