Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178257.2 | NP_839988.1 | 7 | 1318 | ||
GenBank mRNA | AK141289.1 | - | 7 | 1319 | ||
AY103454.1 | - | 7 | 1257 | |||
BC140972.1 | - | 7 | 1324 | 9055584 | ||
BC140973.1 | - | 7 | 1324 | 9055585 |