Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284270.1 | NP_001271199.1 | 4, 5 | 736 | ||
NM_001284273.1 | NP_001271202.1 | 4, 5 | 723 | |||
NM_177672.4 | NP_808340.2 | 4, 5 | 736 | |||
GenBank mRNA | AK040555.1 | - | 1, 2 | 1161 | ||
AK078532.1 | - | 4, 5 | 663 | |||
AK079157.1 | - | 4, 5 | 643 | |||
AK153631.1 | - | 4, 5 | 713 | |||
AK159041.1 | - | 4, 5 | 724 | |||
BC072645.1 | - | 4, 5 | 702 | 30357476 | ||
CA316211.1 | - | 2, 3 | 361 | |||
CD353644.1 | - | 4, 5 | 604 |