Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144908.3 | NP_659157.1 | 2 | 440 | ||
GenBank mRNA | AF495528.1 | - | 1 | 312 | ||
AK088014.1 | - | 2 | 429 | |||
AK161524.1 | - | 2 | 350 | |||
BC011428.1 | - | 2 | 416 | 2631696 | ||
BC021504.1 | - | 2 | 441 | 5346678 | ||
BC031150.1 | - | 1 | 340 | |||
BC034184.1 | - | 1 | 340 | 4974216 | ||
BC034185.1 | - | 1 | 392 | 4974695 | ||
BC036143.1 | - | 1 | 341 | 4974645 | ||
BC036145.1 | - | 1 | 334 | 4975474 | ||
Y12435.1 | - | 1 | 263 |