Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048192.2 | NP_001041657.1 | 9, 10 | 1735 | ||
NM_001294262.1 | NP_001281191.1 | 8, 9 | 1763 | |||
NM_174849.3 | NP_777274.1 | 8, 9 | 1652 | |||
GenBank mRNA | AK029676.1 | - | 8, 9 | 1653 | ||
AK034894.1 | - | 8, 9 | 1741 | |||
AK035680.1 | - | 6, 7 | 1220 | |||
BC057349.1 | - | 8, 9 | 1653 | 6813901 | ||
DQ867034.1 | - | 8, 9 | 1648 | |||
DQ867035.1 | - | 8, 9 | 1648 | |||
DQ867036.1 | - | 9, 10 | 1735 | |||
DQ867037.1 | - | 8, 9 | 1648 | |||
DQ867038.1 | - | 8, 9 | 1648 |