Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139064.2 | NP_620703.1 | 5 | 1016 | ||
GenBank mRNA | AJ304865.1 | - | 5 | 1016 | ||
AK160165.1 | - | 5 | 974 | |||
BC052083.1 | - | 6 | 1062 | 6404610 |