Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098476.2 | NP_001091946.1 | 9 | 1149 | ||
NM_178700.5 | NP_848815.2 | 9 | 1477 | |||
GenBank mRNA | AK044889.1 | - | 9 | 1478 | ||
AK045986.1 | - | 9 | 1431 | |||
AK077782.1 | - | 9 | 1478 | |||
AK089014.1 | - | 8 | 1013 | |||
AK168385.1 | - | 9 | 1121 | |||
AK168459.1 | - | 9 | 1121 |