Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285511.1 | NP_001272440.1 | 11 | 1499 | ||
NM_001285514.1 | NP_001272443.1 | 11 | 1373 | |||
NM_144910.2 | NP_659159.1 | 11 | 1367 | |||
NM_178854.4 | NP_849185.2 | 11 | 1377 | |||
GenBank mRNA | AK030112.1 | - | 11 | 1375 | ||
BC018506.1 | - | 11 | 1367 | 3598248 | ||
BC023923.1 | - | 7 | 889 |