Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115010.1 | NP_001108482.1 | 7 | 1724 | ||
NM_172714.4 | NP_766302.2 | 7 | 1592 | |||
GenBank mRNA | AK029288.1 | - | 8 | 1038 | ||
AK030894.1 | - | 7 | 1726 | |||
AK134320.1 | - | 7 | 1590 | |||
AK220225.1 | - | 7 | 1587 | |||
BC106126.1 | - | 6 | 1228 | |||
BC125580.1 | - | 5 | 719 | |||
BC145025.1 | - | 5 | 719 | 9053573 |