Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A830010M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007574.2 | NP_001007575.2 | 1 | 28 | ||
GenBank mRNA | AK043587.1 | - | 1 | 29 |