Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145209.3 | NP_660210.1 | 7 | 1545 | ||
GenBank mRNA | AB067533.1 | - | 6 | 1470 | ||
AK078690.1 | - | 7 | 1546 | |||
AK089111.1 | - | 6 | 1517 | |||
AK149824.1 | - | 6 | 1469 | |||
AK150863.1 | - | 6 | 1599 | |||
AK165578.1 | - | 6 | 1582 | |||
AK172183.1 | - | 2 | 1969 | |||
AY057107.1 | - | 6 | 1458 | |||
AY089728.1 | - | 6 | 1470 |