Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145209.3 | NP_660210.1 | 4 | 602 | ||
GenBank mRNA | AB067533.1 | - | 3 | 527 | ||
AF426289.1 | - | 3 | 533 | |||
AK078690.1 | - | 4 | 603 | |||
AK089111.1 | - | 3 | 572 | |||
AK149824.1 | - | 3 | 526 | |||
AK150863.1 | - | 3 | 656 | |||
AK165578.1 | - | 3 | 639 | |||
AY057107.1 | - | 3 | 515 | |||
AY089728.1 | - | 3 | 527 | |||
BC032152.1 | - | 3 | 539 | 5068699 |