Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145564.3 | NP_663539.1 | 10 | 1445 | ||
GenBank mRNA | AB093270.1 | - | 10 | 1410 | ||
AK048131.1 | - | 10 | 1403 | |||
AK048542.1 | - | 10 | 1403 | |||
AK049723.1 | - | 10 | 1446 | |||
BC021871.1 | - | 10 | 1482 | 3709413 | ||
BC026383.1 | - | 1 | 58 |