Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163470.1 | NP_001156942.1 | 2 | 126 | ||
NM_172275.2 | NP_758479.2 | 2 | 126 | |||
GenBank mRNA | AK150038.1 | - | 2 | 95 | ||
AK192786.1 | - | 1 | 49 | |||
BC038396.1 | - | 2 | 82 | 3987227 | ||
BU613615.1 | - | 2 | 126 |